missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QARS Polyclonal Antibody

Descrizione
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (91%), Chicken (91%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family.
Specifica
Specifica
| Antigene | QARS |
| Applicazioni | Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.64 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | QARS1 |
| N. accesso geni | P47897 |
| Alias gene | 1110018N24Rik; 1200016L19Rik; C80286; GLNRS; glutamine-tRNA ligase; Glutamine--tRNA ligase; glutamine-tRNA synthetase; glutaminyl-tRNA synthetase; MSCCA; PRO2195; QARS; RGD1562301 |
| Simboli geni | QARS1 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?