Learn More
Swann-Morton™ Qlicksmart SnapIT Trolley, Large, Red Ampule Opener
Product Code. 15681464
Change view
Click to view available options
Color:
Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15681464 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15681464 Supplier Swann-Morton™ Supplier No. TE01L
  • ONLINE EXCLUSIVE PRICE
    78.75€
    75.09€ / pezzi 1
    Risparmia 3.66€
    4% Sconto
Spedizione stimata: 17-12-2026
Safer way to open glass ampules and avoid painful and dangerous sharps injuries.
  • This easy-to-use multi-use ampule opener helps to avoid unacceptable sharps injuries rates seen when ampules are opened by hand.
  • Opening ampules is a particularly high-risk event with 26% of needlestick and sharps injuries (NSIs) being caused by opening an ampule or by broken ampules.
  • The device was invented by a registered nurse in Rockhampton, Qld, Australia who recognized that millions of tamperproof glass ampules are used each year within health care and related practice to contain pharmaceuticals.
  • The practical benefits are clear however the injury rates have always been significant.
  • You simply insert the ampule lid into the multi use SnapIT making sure you have the correct size.
  • If there is a dot on the ampule then align this with the dot on the SnapIT.
  • Snap away from your body to open the ampule.
  • You can then eject the glass ampule lid directly into a sharps container.
  • There are both personal and trolley versions available in the range:
    • Trolley Edition: for team use featuring a flat side so it can lay on a bench top or trolley without rolling away - perfect for hospital medication preparation areas.
    • Pocket Edition: anodized aluminum for personal use on a key ring, hung from a lanyard or kept in your pocket.
    • Lite Edition: made from sturdy, low cost plastic.
  • Regular size Snap-It can be used with ampules of 1-2 mL, 5-10 mL and 10-15 mL and the large trolley edition can be used with ampules of 5-10 mL and 20-25 mL.
Specifications
Color Red
Product Type Ampule Opener
Material Anodized Aluminum
Size Large