Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Shop All Thermo Scientific ProductsDescription
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
Reduces Labor
- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
- Liquid stable and ready-to-use
- Applicable to general chemistry analyzers
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Low cross-reactivity with metabolites and commonly co-administered drugs
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Amikacin |
| Sample Type | Plasma, Serum |
| Type | Calibrator |
| Quantity | Levels A-F, 1 x 1 mL |