Learn More
Thermo Scientific™ QSP PCR Tubes and Plates
Thermo Scientific™ QSP PCR Tubes and Plates are molded from 100% virgin polypropylene.
dietro preventivo
Specifica
Certificazione/Conformità Free of Rnase, Dnase, DNA, Pyrogen
Colore Natural
Descrizione QSP 0.2ML
Da utilizzare con (apparecchiatura) Thermal Cyclers
Sterilità Non-sterile
Visualizza altre specifiche
Descrizione
Descrizione
Thermo Scientific™ QSP PCR Tubes are certified RNase/DNase, DNA, and Pyrogen free.  Packaged in non sterile, bulk bags. Available in colors: red, blue, green, yellow, orange and violet.
  • Tubes feature thin walls for optimal heat transfer
  • High clarity provides sample visibility
  • Caps seal securely to minimize evaporation
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile
Specifica
Specifica
Free of Rnase, Dnase, DNA, Pyrogen
QSP 0.2ML
Non-sterile
Natural
Thermal Cyclers