Learn More
Macherey Nagel Qualitative Analysis Test Papers
For the qualitative determination of ions and chemical compounds.
Marca: Macherey Nagel 90755
Descrizione
- Test papers allow the qualitative determination of ions and chemical compounds
- They are used to detect whether compounds tested for are present above the specific detection limit
- Some of the papers have specific applications
Specifica
| Nitrite ions, nitrous acid, ozone | |
| Refill pack of 3 rolls | |
| Qualitative analysis test strips |