Learn More
R&D Systems™ Quantikine Immunoassay Control Group QC310
Product Code. 18715915
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18715915 1 Ea. 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18715915 Supplier R&D Systems™ Supplier No. QC310
  • ONLINE EXCLUSIVE PRICE
    266.72€
    238.35€ / 1 sets
    Risparmia 28.37€
    10% Sconto
 Restricted Item
Spedizione stimata: 19-11-2026
Controls for Human IL-1 beta/IL-1F2 Quantikine ELISA
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Gene ID (Entrez) 3553
Product Line R&D Systems
Quantity 1 Ea.