Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1149 for Human Hepcidin
Specifica
Specifica
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Tipo | Quantikine Immunoassay Control Set 881 for Human Hepcidin |
| Intended Use | Control |
| Linea di prodotti | R&D Systems |
| Quantità | 1 Each |