Learn More
R&D Systems™ Quantikine Immunoassay Control Set 985 for Human MIF
Product Code. 18796709
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796709 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18796709 Supplier R&D Systems™ Supplier No. QC122
  • ONLINE EXCLUSIVE PRICE
    266.72€
    238.35€ / pezzi 1
    Risparmia 28.37€
    10% Sconto
 Restricted Item
Spedizione stimata: 27-11-2026
For use with catalog numbers DMF00B, SMF00B, PDMF00B
Quantikine Immunoassay Control Set 985 for human MIF is designed for use as quantitative controls for the determination of cytokine concentrations in biological fluids.
  • Analyte: Human Macrophage Migration Inhibitory Factor (MIF)
  • Format: Lyophilized
  • Content: Recombinant human MIF at low, medium, and high concentrations in a buffered protein solution with preservatives
  • Applications: ELISA control
  • Storage: 2°C to 8°C (unreconstituted)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes MIF
Sample Type Plasma
Type Immunoassay Control Set
Gene ID (Entrez) 4282
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized