missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vilber Quantum CX5 with blue transiluminator and EU power cord
Descrizione
- High-Quality Imaging: Scientific-grade camera made in Germany ensures precise detection and high-resolution images for accurate analysis.
- Exceptional Sensitivity: Detects very faint DNA, RNA, or protein bands, enabling reliable results even for low-abundance samples.
- Publication-Ready Results: Combines sensitivity and resolution to deliver TIFF images suitable for scientific publications.
- Clear Band Separation: Advanced imaging technology allows clear distinction of closely spaced bands.
- Separated PC: Users have full freedom to install any software or security they require on the PC controlling the Quantum.
- Durable Construction: Steel and stainless steel darkroom designed for long-lasting performance and easy cleaning.
- Advanced Camera Features: 5 Megapixels extendable to 20, passive cooling, USB3 connection, 16-bit imaging with 65,536 grey levels, and motorized zoom lens.
- Complete and Practical: Small footprint suitable for lab benches, full network connectivity, 3D Dynamic Scan, and BioVision software for advanced image editing and analysis.
- Time-Saving: Automatic functions and intuitive interface reduce hands-on time and simplify experiments.
- Safe and Reliable: Automatic blue shut-off protects users and samples.
- Versatile Applications: Suitable for DNA, RNA, and protein gels in research, teaching, and diagnostic laboratories.
Specifica
Specifica
| Include | EU Power Cord |
| Da utilizzare con (applicazione) | Gel Documentation |
| Product Type | Gel Documentation System |
| Tipo di display | None |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?