missing translation for 'onlineSavingsMsg'
Learn More

Quercetin 3-O-(6' '-O-Malonyl)-b-D-glucoside, TRC

Codice prodotto. 30419237
Cambia vista
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30419237 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30419237 Fornitore Toronto Research Chemicals N. del fornitore TRCQ51003510MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Materiale o nome chimico Quercetin 3-O-(6''-O-Malonyl)-b-D-glucoside
CAS 96862-01-0
Formula molecolare C24 H22 O15
Quantità 10 mg
Sinonimo 3-[[6-O-(2-Carboxyacetyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Quercetin 3-O-(6-O-malonyl)-β-D-glucoside
InChI Formula InChI=1S/C24H22O15/c25-9-4-12(28)17-13(5-9)37-22(8-1-2-10(26)11(27)3-8)23(19(17)33)39-24-21(35)20(34)18(32)14(38-24)7-36-16(31)6-15(29)30/h1-5,14,18,20-21,24-28,32,34-35H,6-7H2,(H,29,30)/t14-,18-,20+,21-,24+/m1/s1
SMILES O[C@H]1[C@H](O)[C@@H](COC(=O)CC(=O)O)O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@@H]1O
IUPAC Name 3-[[(2R,3S,4S,5R,6S)-6-[2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4-oxochromen-3-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3-oxopropanoic acid
Molecular Weight (g/mol) 550.423
Peso formulazione 550.096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.