Learn More
Condensatore a serpentina in vetro borosilicato Quickfit™
Descrizione
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
- Coolant is circulated through a coil having a very large cooling surface
- Ideal for use in the upright reflux mode where counter-current cooling is possible
- Supplied with detachable plastic connectors for the attachment of rubber hose
Specifica
Specifica
| Da utilizzare con (applicazione) | Ideale per l'uso in modalità di riflusso verticale quando è possibile il raffreddamento controcorrente |
| Lunghezza camicia (sistema metrico) | 320 mm (Overall) 160 mm (Effective) |
| Nessun giunto rastremato standard | 34/35 |
| Materiale | Vetro borosilicato Pyrex |
| Tipo | Spirale per condensatore |