Learn More
Quickfit™ Dropping Funnel
Product Code. 15728037
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15728037 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15728037 Supplier Quickfit Supplier No. ML/FUN
  • ONLINE EXCLUSIVE PRICE
    68.62€
    57.30€ / pezzi 1
    Risparmia 11.32€
    16% Sconto
Spedizione stimata: 04-03-2027
Quickfit™ Dropping Funnel is designed for use with the ML1 microscale kit.
  • Interchangeable PTFE stopcock key
  • Supplied complete with screwthread connector, O ring and sealing disc
  • Can be used in conjunction with microscale glass stopper
Specifications
Capacity (Metric) 25 mL
For Use With (Equipment) ML1 microscale kit
Includes Interchangeable PTFE stopcock key
Product Type Dropping Funnel