Learn More
(R)-3-(Boc-amino)piperidine, 97%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar H52749.14
| Quantità | 25g |
|---|
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifica
| (R)-3-(Boc-amino)piperidine | |
| C10H20N2O2 | |
| 25g | |
| WUOQXNWMYLFAHT-MRVPVSSYSA-N | |
| tert-butyl N-[(3R)-piperidin-3-yl]carbamate | |
| 1514172 | |
| 97% |
| 309956-78-3 | |
| MFCD03093382 | |
| r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester | |
| CC(C)(C)OC(=O)NC1CCCNC1 | |
| 200.282 | |
| 200.28 |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.