Learn More
R&D Systems™ Canine CD45 (XP_005622335) VersaClone cDNA
Codice prodotto. 18730648
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18730648 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18730648 Fornitore R&D Systems™ N. del fornitore RDC1145
Please to purchase this item. Need a web account? Register with us today!
Cluster of Differentiation 45
The Canine CD45 (XP_005622335) VersaClone cDNA clone contains the complete ORF for CD45, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD45 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifica
Gene CD45/PTPRC
Numero di accesso XP_005622335
ID gene (immissione) 5788
Alias gene B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specie Canine
Quantità 10 μg
For Reasearch Use Only; Not for Use in Humans