missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40/TNFRSF5 (NP_001241) VersaClone cDNA

Codice prodotto. 18735679
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18735679 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18735679 Fornitore R&D Systems™ N. del fornitore RDC1325

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CD40 type II isoform; CD40L receptor

The Human CD40/TNFRSF5 (NP_001241) VersaClone cDNA clone contains the complete ORF for CD40/TNFRSF5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD40/TNFRSF5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifica

Product Type cDNA
Descrizione Gene ID (Entrez): 958, 21939, 171369
Gene CD40/TNFRSF5
Numero di accesso NP_001241
Concentrazione 10 μg at 0.2 μg/μL
Formulazione cDNA is provided in 10mM Tris-Cl, pH 8.5
Tipo di inserto 847bp
ID gene (immissione) 958
Alias gene B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specie Human, Mouse, Rat
Quantità 10 μg
Vedi altri risultati Mostra meno risultati

For Research Use Only; Not for Use in Humans

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.