Learn More
R&D Systems™ Human GPR148 (NP_997247) VersaClone cDNA
Descrizione
The Human GPR148 (NP_997247) VersaClone cDNA clone contains the complete ORF for GPR148, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR148 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Gene | hGPR148 |
| Numero di accesso | NP_997247 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 1057bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |
For Reasearch Use Only; Not for Use in Humans