Learn More
R&D Systems™ MagCellect Mouse B Cell Isolation Kit
Codice prodotto. 18472480
Cambia vista
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18472480 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18472480 Fornitore R&D Systems™ N. del fornitore MAGM204
  • ONLINE EXCLUSIVE PRICE
    823.00€
    778.05€ / 1 sets
    Risparmia 44.95€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 03-08-2026
Aggiungi al carrello
Aggiungi al carrello
Designed to isolate B cells via a negative selection principle
Typical recovery ranges between 30% and 55% and the purity of recovered B cells ranges from 90-97%
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction), leaving the untagged cells or desired cell population in suspension to be harvested by aspiration while the tube remains in the magnetic field
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantità 1 Kit