Learn More
R&D Systems™ MycoProbe Mycoplasma Detection Kit
Product Code. 18791954
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18791954 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18791954 Supplier R&D Systems™ Supplier No. CUL001B
  • ONLINE EXCLUSIVE PRICE
    743.15€
    661.50€ / 1 sets
    Risparmia 81.65€
    10% Sconto
 Restricted Item
Spedizione stimata: 25-11-2026
A complete kit to detect common antibiotic-resistant cell culture contaminants
Mycoplasma contamination affects up to 80% of continuous cell cultures. If undetected, mycoplasma contamination can have significant effects on the quality and reliability of cell culture preparations.
Reduces experimental variation
Accurate and highly sensitive
Simple multiwell-based colorimetric assay
Only takes 4.5 hours
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit