Learn More
R&D Systems™ Proteome Profiler Human Apoptosis Array Kit
Codice prodotto. 18047621
Cambia vista
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18047621 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18047621 Fornitore R&D Systems™ N. del fornitore ARY009
  • ONLINE EXCLUSIVE PRICE
    822.00€
    777.00€ / 1 sets
    Risparmia 45.00€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 07-10-2026
The Proteome Profiler Human Apoptosis Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Apoptosis Array Kit contains 4 membranes - each spotted in duplicate with 35 different apoptosis antibodies.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantità 1 Kit