Learn More
R&D Systems™ Recombinant Human MMP-1 Western Blot Standard Protein
Codice prodotto. 18327757
Cambia vista
Click to view available options
Quantità:
1 Vial
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18327757 1 Vial 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18327757 Fornitore R&D Systems™ N. del fornitore WBC024
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
The Recombinant Human MMP-1 Western Blot Standard Protein has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) Western Blot
ID gene (immissione) 4312
Nome MMP-1 Western Blot Standard
Quantità 1 Vial
Requisiti di stoccaggio Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Simbolo del gene MMP1
Coniugato Unconjugated
Recombinant Recombinant
Proteine MMP-1