Learn More
R&D Systems™ StemXVivo Chondrogenic Base Media
Codice prodotto. 18472130
Cambia vista
Click to view available options
Quantità:
50 mL
Dimensione della confezione:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18472130 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18472130 Fornitore R&D Systems™ N. del fornitore CCM005
  • ONLINE EXCLUSIVE PRICE
    262.00€
    247.80€ / 50mL
    Risparmia 14.20€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 21-08-2026
Aggiungi al carrello
Aggiungi al carrello
Base media for the differentiation of MSCs into chondrocytes
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce chondrogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Human/Mouse or Rat Chondrogenic Supplements to reduce variation during chondrogenesis
TRUSTED_SUSTAINABILITY
Specifica
Product Type Stem Cell Expansion Media
Quantità 50 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.