Learn More
R&D Systems™ Streptavidin-HRP
Product Code. 18452110
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18452110 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18452110 Supplier R&D Systems™ Supplier No. 48003006
  • ONLINE EXCLUSIVE PRICE
    162.88€
    144.90€ / 30µL
    Risparmia 17.98€
    11% Sconto
 Restricted Item
Spedizione stimata: 20-11-2026
Provided in liquid form with a proprietary stabilizer added, without preservative
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 μL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Type Streptavidin-HRP
Product Line R&D Systems