Learn More
(R)-Norfluoxetine Phthalimide, TRC
Product Code. 30419467
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30419467 Supplier Toronto Research Chemicals Supplier No. TRCN68114050MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-Norfluoxetine Phthalimide
CAS 878663-12-8
Molecular Formula C24H18F3NO3
Quantity 50 mg
Synonym 2-[(3R)-3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-1H-isoindole-1,3(2H)-dione, (R)-1-(Phthalimido)-3-[(4-trifluoromethylphenyl)oxy]-3-phenylpropane
InChI Formula InChI=1S/C24H18F3NO3/c25-24(26,27)17-10-12-18(13-11-17)31-21(16-6-2-1-3-7-16)14-15-28-22(29)19-8-4-5-9-20(19)23(28)30/h1-13,21H,14-15H2/t21-/m1/s1
SMILES FC(F)(F)c1ccc(O[C@H](CCN2C(=O)c3ccccc3C2=O)c4ccccc4)cc1
IUPAC Name 2-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]isoindole-1,3-dione
Molecular Weight (g/mol) 425.4
Formula Weight 425.1239