Learn More
R&D Systems™ Quantikine Immunoassay Control Set 653 for Human Galectin-1
Product Code. 30001924
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001924 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001924 Supplier R&D Systems™ Supplier No. QC94
 Restricted Item
Spedizione stimata: 01-12-2026
For use with catalog number DGAL10
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Galectin-1
Gene ID (Entrez) 3956
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human