Learn More
(R,S)-N-Nitroso Anabasine-d4 (0.1 mg/mL in Methanol), TRC
Specifications
Specifications
| Materiale o nome chimico | (RS)-N-Nitrosoanabasine pyridinyl-d4 |
| CAS | 1020719-68-9 |
| Formula molecolare | C10 2H4 H9 N3 O |
| Quantità | 5x1 mL |
| Sinonimo | Pyridine-2,3,4,6-d4, 5-(1-nitroso-2-piperidinyl)-, 5-(1-Nitroso-2-piperidinyl)pyridine-2,3,4,6-d4, (R,S)-N-Nitroso Anabasine-d4, (RS)-N-Nitrosoanabasine D4 (pyridinyl D4), (RS)-N-Nitrosoanabasine D4 (pyridinyl D4) |
| InChI Formula | InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/i3D,4D,6D,8D |
| SMILES | [2H]c1nc([2H])c(C2CCCCN2N=O)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine |
| Molecular Weight (g/mol) | 195.25 |
| Peso formulazione | 195.131 |
| Show More |