Learn More
RAB32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA555249
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: NKDSSQSPSQ VDQFCKEHGF AGWFETSAKD NINIEEAARF LVEKILVNHQ SFPNEENDVD KIKLDQETLR AENKSQCC Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 72%.
RAB32 Polyclonal antibody specifically detects RAB32 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
RAB32 | |
Polyclonal | |
Unconjugated | |
RAB32 | |
2810011A17Rik; AU022057; RAB32; RAB32, member RAS oncogene family; ras-related protein Rab-32; RGD1559997 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10981 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13637 | |
RAB32 | |
Recombinant protein corresponding to Human RAB32 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only