Learn More
COL5A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descrizione
This gene encodes an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This gene product is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations. Mutations in this gene are associated with Ehlers-Danlos syndrome, types I and II. [provided by RefSeq
Specifica
Specifica
| Antigene | COL5A1 |
| Applicazioni | Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Descrizione | Rabbit polyclonal antibody raised against synthetic peptide of COL5A1. |
| Diluizione | Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulazione | In PBS, pH 7.2 (0.05% sodium azide) |
| Gene | COL5A1 |
| Simboli geni | COL5A1 |
| Vedi altri risultati |
For Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.