missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 550
Descrizione
This antibody is cross-adsorbed and exhibits minimum reactivity to mouse and rat.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
Specifica
| Antigene | Human IgG (H+L) Cross-Adsorbed |
| Applicazioni | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classificazione | Polyclonal |
| Concentrazione | 0.5 mg/mL |
| Coniugato | DyLight 550 |
| Formulazione | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Specie ospite | Rabbit |
| Immunogeno | Human IgG-heavy and light chain. |
| Metodo di purificazione | Antigen affinity chromatography |
| Quantità | 500 μg |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?