missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680

Codice prodotto. 13437317
Cambia vista
Click to view available options
Quantità:
500 μg
Dimensione della confezione:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
13437317 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 13437317 Fornitore Invitrogen™ N. del fornitore SA510162

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Secondary Antibody

This antibody is cross-adsorbed and exhibits minimum reactivity to human and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Mouse IgG (H+L) Cross-Adsorbed
Applicazioni Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 0.5 mg/mL
Coniugato DyLight 680
Formulazione PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4
Specie ospite Rabbit
Immunogeno Mouse IgG-heavy and light chain.
Metodo di purificazione Antigen affinity chromatography
Quantità 500 μg
Status giuridico RUO
Primario o secondario Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.