missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLE1, Rabbit, Polyclonal Antibody, Abnova™
Description
Sequence: TIKVWRAHDGVLCRTLQGHGHWVNTMALSTDYALRTGAFEPAEASVNPQDLQGSLQELKERALSRYNLVRGQGPERLVSGSDDFTL
Specifications
Specifications
| Antigene | NLE1 |
| Applicazioni | Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | Rabbit polyclonal antibody raised against recombinant NLE1. |
| Diluizione | Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulazione | In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) |
| Gene | NLE1 |
| Alias gene | FLJ10458/Nle |
| Simboli geni | NLE1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?