Learn More
Invitrogen™ Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680
Codice prodotto. 13497207
Cambia vista
Click to view available options
Quantità:
500 μg
Dimensione della confezione:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
13497207 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 13497207 Fornitore Invitrogen™ N. del fornitore SA510058
Rabbit Polyclonal Secondary Antibody
This antibody is cross-adsorbed and exhibits minimum reactivity to chicken, horse, human, mouse and rat.
Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifica
Antigene Sheep IgG (H+L) Cross-Adsorbed
Applicazioni Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 0.5 mg/mL
Coniugato DyLight 680
Formulazione PBS with 0.09% sodium azide; pH 6.8 to 7.4
Specie ospite Rabbit
Immunogeno Sheep IgG-heavy and light chain.
Metodo di purificazione Antigen affinity chromatography
Quantità 500 μg
Status giuridico RUO
Primario o secondario Secondary
Target Species Sheep
Content And Storage 4°C
Product Type Secondary Antibody
Forma Liquid
Isotype IgG