Learn More
UTS2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descrizione
This gene encodes a mature peptide that is an active cyclic heptapeptide absolutely conserved from lamprey to human. The active peptide acts as a vasoconstrictor and is expressed only in brain tissue. Despite the gene family name similarity, this gene is not homologous to urocortin, a member of the sauvagine/corticotropin-releasing factor/urotensin I family. Most of the proprotein is cleaved to make the mature peptide. Transcript variants encoding different preproprotein isoforms have been described for this gene. [provided by RefSeq
Specifica
Specifica
| Antigene | UTS2 |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Descrizione | Rabbit polyclonal antibody raised against synthetic peptide of UTS2. |
| Diluizione | Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. |
| Formulazione | In PBS, pH 7.2 (0.05% sodium azide) |
| Gene | UTS2 |
| Alias gene | PRO1068/U-II/UCN2/UII |
| Vedi altri risultati |
For Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.