missing translation for 'onlineSavingsMsg'
Learn More

VASP (phospho S238), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Codice prodotto. 16085997
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16085997 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16085997 Fornitore Abnova N. del fornitore PAB25820.100ug

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit polyclonal antibody raised against synthetic phosphopeptide of VASP.

Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG. [provided by RefSeq

Sequence: K-V-Sp-K-Q

Specifica

Antigene VASP
Applicazioni Immunohistochemistry (PFA fixed), Western Blot
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Descrizione Rabbit polyclonal antibody raised against synthetic phosphopeptide of VASP.
Diluizione Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulazione In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene VASP
Simboli geni VASP
Specie ospite Rabbit
Immunogeno Synthetic phosphopeptide corresponding to residues surrounding S238 of human VASP.
Metodo di purificazione Affinity chromatography
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 7408
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Forma Liquid
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.