Descrizione
The RABL4 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifica
Specifica
| Linea cellulare | The lysate was created in HEK293T cells, using plasmid ID RC2215 and based on accession number NM_686. The protein contains a C-terminal DDK tag. |
| Da utilizzare con (applicazione) | Western Blot |
| Quantità | 0.1 mg |
| Tampone di conservazione | RIPA buffer |
| Purezza | Protein |
| Test di specificità | Homo sapiens RAB, member of RAS oncogene family-like 4 (RABL4), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only