missing translation for 'onlineSavingsMsg'
Learn More

Rabusertib, MedChemExpress

Codice prodotto. 16498737
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 16498737

Marca: Medchem Express HY14720

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM.

Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM.

Specifications

Materiale o nome chimico Rabusertib
CAS 911222-45-2
Colore White
Note sul grado di purezza Research
Formula molecolare C18H22BrN5O3
Quantità 5 mg
Sinonimo LY2603618 IC-83
Informazioni di solubilità DMSO : 31.25 mg/mL (71.63 mM; ultrasonic and warming and heat to 60°C)
SMILES BrC(C=C1NC(NC2=NC=C(N=C2)C)=O)=C(C=C1OC[C@H]3OCCNC3)C
Molecular Weight (g/mol) 436.3
Peso formulazione 436.3
Percent Purity 97.05%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.