Learn More
rac Metanephrine Hydrochloride Salt (0.1mg/mL in Methanol), TRC
Specifica
Specifica
| Materiale o nome chimico | (±)-Metanephrine Hydrochloride |
| CAS | 881-95-8 |
| Formula molecolare | C10H16ClNO3 |
| Quantità | 10 x 1 mL |
| Sinonimo | Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride (1:1), Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride (9CI), Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride, (±)-, Vanillyl alcohol, α-[(methylamino)methyl]-, hydrochloride, (±)- (8CI), (±)-Metanephrine hydrochloride, Metanephrine hydrochloride, dl-Metanephrine hydrochloride, D,L-Metanephrine Hydrochloride, rac-Metanephrine Hydrochloride |
| InChI Formula | InChI=1S/C10H15NO3.ClH/c1-11-6-9(13)7-3-4-8(12)10(5-7)14-2;/h3-5,9,11-13H,6H2,1-2H3;1H |
| SMILES | Cl.CNCC(O)c1ccc(O)c(OC)c1 |
| IUPAC Name | 4-[1-hydroxy-2-(methylamino)ethyl]-2-methoxyphenol;hydrochloride |
| Molecular Weight (g/mol) | 233.69 |
| Peso formulazione | 233.0819 |
| Vedi altri risultati |