Learn More
Hamilton™ Filtered Low Retention Pre-Sterilized Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Marca: Hamilton™ 68007-23
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifica
10 μL | |
Sterile | |
Polypropylene |
Filtered | |
Rack |