Learn More
Ranolazine Dihydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Ranolazine Dihydrochloride |
| CAS | 95635-56-6 |
| Formula molecolare | C24H35Cl2N3O4 |
| Quantità | 250 mg |
| Sinonimo | N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-1-piperazineacetamide Dihydrochloride, (±)-N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-1-piperazineacetamide Dihydrochloride, (±)-RS 43285-193, RS 43285, RS 43285-193, Ranolazine Hydrochloride |
| InChI Formula | InChI=1S/C24H33N3O4.2ClH/c1-18-7-6-8-19(2)24(18)25-23(29)16-27-13-11-26(12-14-27)15-20(28)17-31-22-10-5-4-9-21(22)30-3;;/h4-10,20,28H,11-17H2,1-3H3,(H,25,29);2*1H |
| SMILES | Cl.Cl.COc1ccccc1OCC(O)CN2CCN(CC(=O)Nc3c(C)cccc3C)CC2 |
| IUPAC Name | N-(2,6-dimethylphenyl)-2-[4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl]acetamide;dihydrochloride |
| Molecular Weight (g/mol) | 500.46 |
| Peso formulazione | 499.2005 |