missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAP2A/RAP2B Monoclonal Antibody (455410)
GREENER_CHOICE

Codice prodotto. 30534666
Cambia vista
Click to view available options
Quantità:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30534666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30534666 Fornitore Invitrogen™ N. del fornitore MA561476

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

RAP2A and RAP2B are genes encoding small GTPases that belong to the Ras-related protein family. These proteins are involved in various cellular processes, including cell proliferation, differentiation, and migration. RAP2A and RAP2B function as molecular switches, cycling between an active GTP-bound state and an inactive GDP-bound state, thereby regulating signal transduction pathways. They play crucial roles in the organization of the actin cytoskeleton, vesicle trafficking, and cell adhesion. RAP2A and RAP2B are implicated in the regulation of neuronal development and plasticity, and their dysregulation has been associated with neurological disorders. Additionally, abnormal expression or mutations in these genes have been linked to cancer progression, as they can influence tumor cell migration and invasion. Understanding the specific mechanisms and functions of RAP2A and RAP2B is important for elucidating their roles in both normal cellular physiology and disease states. As such, these genes are significant targets in research aimed at developing therapeutic interventions for cancer, neurodegenerative diseases, and other conditions involving cellular signaling abnormalities.
TRUSTED_SUSTAINABILITY

Specifica

Antigene RAP2A/RAP2B
Applicazioni Western Blot
Classificazione Monoclonal
Clone 455410
Concentrazione 0.5 mg/mL
Coniugato Unconjugated
Formulazione PBS with trehalose and no preservative
Specie ospite Mouse
Immunogeno E. coli-derived recombinant human Rap2A/B, Met1-Gln183.
Metodo di purificazione Protein A/G
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Lyophilized
Isotype IgG2b
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.