Learn More
Thermo Scientific™ RapID™ NH System
Descrizione
Identify over 30 taxa including Neisseria, Moraxella, Haemophilus, and related microorganisms in just four hours using the Thermo Scientific™ RapID™ NH System, an identification system based on enzyme technology.
- One-step inoculation—decreased prep time for increased productivity
- Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
- Four-hour incubation*—quicker time-to-result for a faster response
- Common procedure—lessened materials usage saves time and money
- Visible color reactions—reduced subjectivity decreases repeat testing
- Better reporting—more coverage, advanced updates
*Two-hour incubation for RapID SS/u.
A clear plastic tray contains reagent impregnated wells, allowing simultaneous inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Inoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a database, or through the use of a computer-generated Code Compendium (ERIC).
Requires:
RapID Spot Indole, (R8309002); RapID Nitrate A, (R8309003); RapID Nitrate B (R8309004); RapID Inoculation Fluid, (R8325102)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Specifica
| Content And Storage | 2°C to 8°C |
| Descrizione | RapID NH System |
| Marchio CE | Yes |
| Analiti rilevabili | Neisseria, Moraxella, Haemophilus |
| Tipo di imballaggio | 1 Box |
| Quantità | 20 Panels/Kit |