Learn More
Labconco™ RapidVap Vacuum Dry Evaporation System
Codice prodotto. 11761549 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Tensione:
300 V
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione unitSize
11761549 300 V Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11761549 Fornitore Labconco™ N. del fornitore 7900001
 Articolo soggetto a restrizioni
Spedizione stimata: 23-12-2026
Aggiungi al carrello
Aggiungi al carrello
The RapidVap Vacuum Evaporation Systems are ideal for preparation of samples in a variety of applications including drug discovery, agrichemistry, mycology testing and environmental analysis.
The RapidVap Vacuum Evaporation Systems are ideal for preparation of samples in a variety of applications including drug discovery, agrichemistry, mycology testing and environmental analysis. The systems use vacuum along with heat and vortex motion to help speed evaporation and provide an alternative to traditional centrifugal evaporation methods. These systems may be used with a broad range of aggressive chemicals. Heat is supplied by a dry block heating system that, unlike water baths, adds no potential source of contamination and requires no maintenance.
Specifica
Tensione 300 V
Materiale camera PTFE-coated Aluminum
Descrizione 1000-watt dry block heating system