missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGEF1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550781
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total RASGEF1A protein.
RASGEF1A Polyclonal antibody specifically detects RASGEF1A in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
RASGEF1A | |
Polyclonal | |
Unconjugated | |
RASGEF1A | |
6330404M18Rik; AI835194; CG4853; CG4853 gene product; RasGEF domain family member 1A; RasGEF domain family, member 1A; ras-GEF domain-containing family member 1A; Rasgef1a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221002 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N9B8 | |
RASGEF1A | |
Fusion protein corresponding to residues near the C terminal of human RasGEF domain family, member 1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |