missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat AGTR-2 (NP_036626.1) VersaClone cDNA

Codice prodotto. 18768669
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18768669 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18768669 Fornitore R&D Systems™ N. del fornitore RDC2976

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifica

Gene AGTR-2
Numero di accesso NP_036626.1
ID gene (immissione) 186
Alias gene AGTR2, Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2, AT2ATGR2, AT2R, MR x 88, MR x 88 type-2 angiotensin II receptor
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specie Rat
Quantità 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.