missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
- The assay recognizes rat TNF-α. An antibody specific for TNF-α has been pre-coated onto the microwells. The TNF-α protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for TNF-α is added to detect the captured TNF-α protein.
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
- Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
- TNF, as also known as TNF-alpha, or cachectin, is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily.
- It is expressed as a 26 kDa membrane bound protein and is then cleaved by TNF-alpha converting enzyme (TACE) to release the soluble 17 kDa monomer, which forms homotrimers in circulation. It is produced chiefly by activated macrophages, although it can be produced by many other cell types such as CD4+ lymphocytes, NK cells, neutrophils, mast cells, eosinophils, and neurons.
- It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation.
- Rat and human TNF-alpha share 79% amino acid sequence identity.
- Unlike human TNF-alpha, the rat form is glycosylated.
- In rat deficiency of this gene is associated with defects in response to bacterial infection, with defects in forming organized follicular dendritic cell networks and germinal centers, and with a lack of primary B cell follicles.
Specifica
Specifica
| Numero di accesso | NP_036807 |
| Intervallo saggio | 15.6 to 1000 pg/mL |
| Assay Sensitivity | 7.7 pg/mL |
| Certificazione/Conformità | ISO 13485 |
| Metodo di rilevamento | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Tipo di campione | Plasma, Serum |
| Substrato | DAB |
| Target Species | Rat |
| ID gene (immissione) | 24835 |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- 96TEST Human ABCD1 ELISA Kit
- Attenzione
- Lesione oculare grave/irritazione oculare Categoria 1
- Lesione oculare grave/irritazione oculare Categoria 2
- Corrosione/irritazione della pelle Categoria 2
- Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
- H302-Nocivo se ingerito.
- H315-Provoca irritazione cutanea.
- H318-Provoca gravi lesioni oculari.
- H319-Provoca grave irritazione oculare.
- H332-Nocivo se inalato.
- H335-Può irritare le vie respiratorie.
- P201-Procurarsi istruzioni specifiche prima dell'uso.
- P202-Non manipolare prima di avere letto e compreso tutte le avvertenze.
- P234-Conservare soltanto nel contenitore originale.
- P264-Lavare accuratamente dopo l'uso.
- P270-Non mangiare, né bere, né fumare durante l'uso.
- P273-Non disperdere nell'ambiente.
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P301+P310-IN CASO DI INGESTIONE: contattare immediatamente un CENTRO ANTIVELENI/un medico
- P301+P312-IN CASO DI INGESTIONE accompagnata da malessere: contattare un CENTRO ANTIVELENI/medico
- P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
- P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
- P308+P313-IN CASO di esposizione o di possibile esposizione, consultare un medico.
- P330-Sciacquare la bocca.
- P362-Togliere gli indumenti contaminati.
- P405-Conservare sotto chiave.
- P501b-Smaltire il prodotto/recipiente in conformità con le disposizioni locali/regionali/nazionali/internazionali.
- MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?