Learn More
Rbm20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558068
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: ADWGNGRDSW EHSPYARREE ERDPAPWRDN GDDKRDRMDP WAHDRKHHPR QLDKAELDER PEGGRPHREK YPRSGSPNLP HSVSSYKSRE D Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.
Rbm20 Polyclonal antibody specifically detects Rbm20 in Human samples. It is validated for Immunohistochemistry.Specifica
Rbm20 | |
Polyclonal | |
Unconjugated | |
Rbm20 | |
1110018J23Rik; 2010003H22Rik; AI646761; probable RNA-binding protein 20; Rbm20; RNA binding motif protein 20; RNA-binding motif protein 20; RNA-binding protein 20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282996 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T481 | |
Rbm20 | |
Recombinant protein corresponding to Human Rbm20 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only