Descrizione
RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimise the separation of matrix beads by size during packing to produce homogeneous columns. Once the matrix is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for column packing. Packing reservoirs for each column diameter optimise packing conditions.
Specifica
Specifica
| Materiale | Acetal Plastic (POM), Polypropylene, Superpolyxymethylene, Fluoro RubberNitrile Rubber (NBR) |
| Tipo | RC 16 Packing Reservoir |
| Volume (sistema metrico) | 108 mL |
| Quantità | 1 Pack |