Learn More
Electrothermal Inerting Cap
Product Code. 11806772
Change view
Click to view available options
Description:
22mm Inerting cap
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
11806772 22mm Inerting cap Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11806772 Supplier Electrothermal Supplier No. 3663059
  • ONLINE EXCLUSIVE PRICE
    2235.00€
    1624.45€ / pezzi 6
    Risparmia 610.55€
    27% Sconto
Spedizione stimata: 08-01-2027
Accessory for Omni Reaction Stations
  • For use with O6050, OS6100 and OS6250
Specifications
Description 22mm Inerting cap
For Use With (Equipment) STEM Omni Reaction Stations: O6050, OS6100 and OS6250