Learn More
Hanna Instruments™ Kit per test a reagente delle acque
Descrizione
This high quality reagent set is designed to follow the Phenol red method for colorimetric determination of pH. By simply adding 5 drops from the bottle of reagent to the sample, the reaction between the sample and reagent causes a tint in the sample that varies from yellow to red. The intensity of color is determined by a compatible photometer.
- Designed for samples with an expected range of 6.5 to 8.5 pH, for results with a resolution of 0.1 pH
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifica
Specifica
| Analiti | Phenol Red |
| Da utilizzare con (applicazione) | pH determination |
| Da utilizzare con (apparecchiatura) | Compatible Hanna benchtop and portable photometers |
| Include | 1 dropper bottle |
| N. di test | 100 |
| Quantità | 100/conf. |
| Range | 6.5 to 8.5 pH |
| Metodo test | Rosso fenolo |
| Tipo | Kit per reagente |