Learn More
Lenz Laborglasintrumente Receiving Flasks with PUR Coating
Codice prodotto. 11903401
Click to view available options
Volume (sistema metrico):
250 mL
2 L
Dimensione della confezione:
Pezzo
Codice prodotto. 11903401
Marca: Lenz Laborglasintrumente 03064073
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With spherical joint socket S 35
  • With highly transparent PUR coating
  • Autoclavable
  • Material: DURAN™
Specifica
Materiale Duran Glass
Volume (sistema metrico) 2 L