missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Aspartate aminotransferase, mitochondrial Protein
A DNA sequence encoding the Bos taurus (Bovine) Aspartate aminotransferase, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6108-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_777231.1 | |
Aspartate aminotransferase, mitochondrial Protein | |
Research Use Only | |
GOT2 | |
Recombinant Protein | |
E. coli | |
GLAAAASARA SSWWAHVEMG PPDPILGVTE AFKRDTNSKK MNLGVGAYRD DNGKPYVLPS VRKAEAQIAA KNLDKEYLPI AGLAEFCKAS AELALGENNE VLKSGRYVTV QTISGTGALR IGASFLQRFF KFSRDVFLPK PTWGNHTPIF RDAGMQLQSY RYYDPKTCGF DFTGAIEDIS KIPAQSVILL HACAHNPTGV DPRPEQWKEM ATVVKKNNLF AFFDMAYQGF ASGDGNKDAW AVRHFIEQGI NVCLCQSYAK NMGLYGERVG AFTVVCKDAE EAKRVESQLK ILIRPMYSNP PINGARIAST ILTSPDLRKQ WLHEVKGMAD RIISMRTQLV SNLKKEGSSH NWQHIIDQIG MFCYTGLKPE QVERLTKEFS IYMTKDGRIS VAGVTSGNVA YLAHAIHQVT K | |
Greater than 90% as determined by SDS-PAGE. |
286886 | |
10 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |